Tlačiť
Tubing clamps, Kocher (Ochsner) type
Dodávateľ: Karl Hammacher
231-0251EA 149.05 EUR
231-0251
Tubing clamps, Kocher (Ochsner) type
Svorky Svorky trubkové
Stainless steel, straight. Ideal for stopping or controlling liquid flow rates in tubing connections.
  • Very good thermal resistance, autoclavable
  • Corrosion-resistant
  • Antimagnetic