Tlačiť
Aprons, Tyvek® 500, model PA30L0
Dodávateľ: DuPont
Tyvek®
113-0479EA 448.13 EUR
113-0479
Aprons, Tyvek® 500, model PA30L0
Jednorázové oděvy
The apron is made of Tyvek®, a fabric composed of flash spun high density PE. Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres (down to 1 micron in size). Specially designed for use with Tyvek® apparel, this apron offers enhanced protection for body parts that are more exposed to hazardous substances.
  • Shin-length apron with neck and waist ties
  • Ultra-low-linting fabric
  • Antistatic treatment (EN 1149-5) - on both sides
  • No silicone added
Typical applications are pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up.
Length: 108 cm
Certifikácie: Type PB [6-B]; EN 14126; Cat. III; (EU) 2016/425
Balenie: 100 per box, bulk packed.