Tlačiť
Cooling mat
Dodávateľ: J.D. Honigberg International
HONI405
Cooling mat
This cooling mat is suitable for cooling, shipping and cold storage. The mat consists of approximately 1500 individually sealed bags.
  • Can be cut to the desired size simply using scissors
  • Used to surround samples for low-temperature transport
  • Reusable
W×D: 400×1000 mm