Tlačiť
First-aid cabinets, City Serie
Dodávateľ: Söhngen
City
118-1230 118-1233 118-1232 118-1231
First-aid cabinets, City Serie
Pomůcky první pomoci Skříňky první pomoci
Manufactured from high quality sheet metal 0,75 mm thick.
  • High stability and torsion resistance, rounded edges and corners to avoid injuries
  • Doors rounded, hinged and provided with a high quality sealing
  • Each cabinet has a safety cylinder lock and two keys
Heidelberg
One door with locking facility, complete with screened lettering and colour pictograms. The inside partitioning can be extended by one additional shelf, two drawers and one medicine compartment. Tailored printing available.
W×D×H (mm): 302×362×140
Madrid
One door, lockable, two adjustable shelves, door is equipped with three storage compartments and unit is finished with screened lettering and colour pictograms. The inside partitioning can be extended with one medicine compartment, one additional shelf and six drawers as required when ordering.
W×D×H (mm): 452×552×170
Certifikácie: DIN standard