Tlačiť
Protective sleeve for aspirator bottles
Dodávateľ: CETEC
519-4505EA 28.93 EUR
519-4505 519-4503 519-4504 519-4501 519-4502
Protective sleeve for aspirator bottles
Podstavce Support Sleeves
EPDM.
  • Good seal
  • Better all-round protection for aspirator bottles against knocks
For the secure placement of funnels, filters and suction strainers onto aspirator bottles.