Chcete sa opäť prihlásiť?
Chemical resistant gloves, Showa 878
Dodávateľ: Showa
Butyl rubber gloves provide superior resistance to highly corrosive acids and are excellent for handling ketones and esters. Synthetic rubber also provides the highest permeation resistance to gases and water vapours, but does not offer the physical strength of natural rubber.
  • Unlined, with smooth finish and rolled cuffs
  • Lightweight and flexible
  • Impermeable for working in damp or greasy environments
  • Cuff prevents dirt from entering the glove
Thickness: 0,76 mm
Colour: Black
EN 388 (1121), EN 374 (BIK); Cat. III
Certifikace: EN 388 (1121), EN 374 (BIK); Cat. III
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Glove size Farba Dĺžka Hrúbka Balené Pk Číslo v katalógu VWR Množstvo
8 Black 350 mm 0,70 mm 1 pair/pack 1 PAIR 112-3266
9 Black 350 mm 0,70 mm 1 pair/pack 1 PAIR 112-3267
10 Black 350 mm 0,70 mm 1 pair/pack 1 PAIR 112-3268
11 Black 350 mm 0,70 mm 1 pair/pack 1 PAIR 112-3154
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
ŠPECIFIKÁCIE
Model 878
Ambidextrous/hand-specific Hand-specific
Disposable/reusable Reusable
Finish Smooth
Cuff style Beaded
EN norm EN 388 (1121), EN 374 (BIK)
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo