Chcete sa opäť prihlásiť?
General purpose gloves, Resistex Light Grip 2
Dodávateľ: Honeywell Safety Products
These polyester/cotton knitted gloves with PVC dot coating on both sides are ideal for the handling of dry and slippery parts.
  • Cotton and polyester fibre blend ensures outstanding comfort and unique flexibility
  • PVC dots on both sides ensure a better grip and an extended life cycle
  • Ambidextrous
  • White colour enhances the glove's hygiene and cleanliness
Suitable for use in administration, agriculture, automotive manufacturing, building and construction, logistics and transportation.
Certifikace: EN 388 (113X); Cat. II
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Glove size Farba Dĺžka Pk Číslo v katalógu VWR Množstvo
7 White/blue 250 mm 1 PAIR 112-4049
8 White/blue 250 mm 1 PAIR 112-4050
9 White/blue 250 mm 1 PAIR 112-4051
10 White/blue 250 mm 1 PAIR 112-4052
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
ŠPECIFIKÁCIE
Model Resistex Light Grip 2
Ambidextrous/hand-specific Ambidextrous
Disposable/reusable Reusable
Liner material Polyester/cotton
Finish Dot coating on both sides
Gauge 13
Cuff style Knit wrist
EN norm EN 388 (113X)
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo