Tlačiť
Hľadali ste: Homogenizátory na tkáně
28  results were found
SearchResultCount:"28"
Sort Results
Zobraziť zoznam Easy View (Nové)
Ohodnotiť tieto výsledky vyhľadávania
Dodávateľ: VWR Collection
Opis: Borosilicate glass tube, PTFE pestle with stainless steel shaft. Tissue grinders are used for controlled reduction of particle size and homogenisation...
Dodávateľ: SCHERF PRAZISION GMBH
Opis: Borosilicate glass with ground glass pestle.
Dodávateľ: SCHERF PRAZISION GMBH
Opis: These tissue grinders (borosilicate glass, PTFE pestle with glass tube) combine both conical and cylindrical surfaces to reduce tissue and also produc...
Dodávateľ: SCHERF PRAZISION GMBH
Opis: Designed primarily for cellular work where the nucleus remains intact after homogenisation.
Dodávateľ: SCHERF PRAZISION GMBH
Opis: These borosilicate glass tissue grinders combine both conical and cylindrical surfaces to reduce tissue and also produces a uniform homogenate. Constr...
Dodávateľ: SCHERF PRAZISION GMBH
Opis: CS1 tissue grinders are suitable for standard grinding, tissues are ground within a sealed container to minimise the risk of personal contact.
Dodávateľ: SCHERF PRAZISION GMBH
Opis: Pellet pestles with a stainless steel shaft/CTFE tips.
Dodávateľ: SCHERF PRAZISION GMBH
Opis: Glass tube with a PTFE pestle. Tissue grinders with capacity of 5 to 45 ml have radial serrations on the lower portion of the pestle to deliver the ho...
Dodávateľ: SCHERF PRAZISION GMBH
Opis: Potter-Elvehjem with all-glass construction designed for a motor drive using a stirrer adapter.
Dodávateľ: KARTELL
Opis: Homogeniser, Tissue, Potter, Borosilikátové sklo, 15 ml, Vnitř. Ø×V: 16×155 mm
Katalógové číslo: (SCERSP884900-0000)
Dodávateľ: SCHERF PRAZISION GMBH
Opis: The kit contains a variety of the finest microscale tissue grinders suitable for use on tough connective tissue and soft tissue such as brain or liver...
Merná jednotka: 1 * 1 items
Dodávateľ: SPECTRUM EUROPE B.V.
Opis: The stainless steel Bessman tissue pulveriser consists of a two-component mortar with handles and a pestle, specifically designed for pulverising 10 -...
Dodávateľ: DWK LIFE SCIENCES
Opis: Borosilicate glass. Handheld homogeniser with conical surface, which allows initial size reduction, followed by passage through the cylindrical sectio...
Dodávateľ: DWK LIFE SCIENCES
Opis: Mortar in borosilicate glass, PTFE pestle with stainless steel rod. Handheld homogeniser with conical surface, which allows initial size reduction, fo...
Dodávateľ: DWK LIFE SCIENCES
Opis: Mortar in borosilicate glass, PTFE pestle with stainless steel rod. Serrated PTFE pestle disperses homogenate into mortar cylinder more efficiently. P...
Dodávateľ: DELTALAB
Opis: Clearance between Glass vessel and appropriately sized plunger is approximately 200 µm.
Zavolať a opýtať sa na ceny
Zásoby tejto položky sú obmedzené, no môžu sa nachádzať v inom sklade blízko pri vás. Uistite sa prosím, že ste prihlásený/á na tejto lokalite, aby sa mohli zobraziť dostupné zásoby. Ak sa stále zobrazuje zavolajte a potrebujete pomoc, zavolajte nám prosím na číslo +43 1 97002 - 0.
Zásoby tejto položky sú obmedzené, no môžu sa nachádzať v inom sklade blízko pri vás. Uistite sa prosím, že ste prihlásený/á na tejto lokalite, aby sa mohli zobraziť dostupné zásoby. Ak sa stále zobrazuje zavolajte a potrebujete pomoc, zavolajte nám prosím na číslo +43 1 97002 - 0.
Daná chemikália je regulovaná podľa platnej legislatívy a bude vyžadované vyplnenie formulára. Jeho včasným vyplnením urýchlite dodanie produktu.
-Na zakúpenie tejto položky môže byť potrebná dodatočná dokumentácia. Ak to bude potrebné, skontaktuje sa s vami zástupca spoločnosti VWR.
Tento produkt bol zablokovaný vašou organizáciou. Kontaktujte prosím vaše oddelenie nákupu pre získanie viac informácií.
Pôvodný produkt už nie je dostupný. Je dostupná zobrazená náhrada.
Tento produkt už nie je dostupný. Môžu byť k dispozícii alternatívy, ktoré sa dajú vyhľadať pomocou katalógového čísla VWR uvedeného vyššie. Ak potrebujete ďalšiu pomoc, zavolajte prosím na zákaznícky servis spoločnosti VWR na čísle +43 1 97002 - 0.
1 - 16 of 28
no targeter for Bottom