Tlačiť
384-well microplates, clear bottom, Falcon® Optilux
Dodávateľ: Corning Life Sciences
Falcon® Optilux
736-2044EA 838.85 EUR
736-2044 736-2045
384-well microplates, clear bottom, Falcon® Optilux
Mikrodestičky Mikrodestičky pro HTS
PS, tissue culture-treated, sterile, with lid. These flat-bottom 120 µl microplates are suitable for a wide variety of applications, including high throughput screening.
  • Standard microplate footprint meets SBS specification
  • Rounded square well geometry reduces wicking
  • Tissue culture-treated for optimum cell attachment and growth