Tlačiť
Deep well plates, Eppendorf deepwell plate 96 and 384
Dodávateľ: Eppendorf
Deepwell
737-0206EA 1182.69 EUR
737-0206 737-2535 737-2564 737-2514 737-0211 737-2526 737-0210 737-2518 737-2508 737-2519 737-0203 737-2545 737-2505 737-2504 737-2541 737-2540 737-2528 737-2529 737-2516 737-2549 737-0207 737-0209 737-2513 737-2530 737-2511 737-0200 737-2503 737-2500 737-2501 737-0213 737-2534 737-0205 737-2525 737-2509 737-2510 737-2521 737-2506 737-2524 737-2539 737-0212 737-2523 737-2515 737-0208 737-0201 737-2554 737-0161 737-2544 737-0202 737-0204 737-2520 737-2559
Deep well plates, Eppendorf deepwell plate 96 and 384
Mikrodestičky Destičky s hlubokými jamkami
PP. For processing, transport and storage of liquids. The well geometry enables high-speed pipetting, excellent mixing and high-speed centrifugation to accelerate both manual and automated work processes. Colour coding makes reading, orientation and manual handling both easier and faster. Available in standard or sterile quality, Eppendorf Deepwell plate 96/500, 96/1000 and 384/200 are also available as DNA LoBind and Protein LoBind quality. DNA LoBind and Protein LoBind plates are certified PCR clean (free from DNase, RNase, human DNA and PCR inhibitors).
  • Centrifugation stable and durable in a temperature range of +4 to +40 °C
  • Standard plates can be centrifuged up to 6000 ×g; sterile plates up to 5000 ×g
  • High chemical resistance
  • Autoclavable (121 °C, 20 min)
  • Conforms to SBS standards
  • Raised well rims for effective sealing by adhesive seal, heat sealing or sealing mats
Objednávacie informácie: Deepwell plate 384/200 and Deepwell plate 96/500 are supplied in packs of 40 plates (5 bags of 8); Deepwell plate 96/1000 and Deepwell plate 96/2000 are supplied in packs of 20 plates (5 bags of 4).