Tlačiť
Storage plate cap strips
Dodávateľ: Thermo Fisher Scientific
732-4922EA 70.19 EUR
732-4922
Storage plate cap strips
Těsnění Těsnění mikrodestičky
Strips of eight caps for sealing storage plates and racked cluster tubes. The strips are compatible with Thermo Scientific microplates, 0,8 ml storage plates, 1,2 ml round well storage plates and 1,2 ml cluster tubes ranges.
  • Each cap is individually numbered
  • Simultaneous sealing of 8 wells
  • End tabs for ease of removal
  • Low profile for increased well volume