Tlačiť
Pipette stand
Dodávateľ: Eppendorf
612-7489EA 7.33 EUR
EPPE4980210.001
Pipette stand
Carousel rack with small footprint for six Eppendorf® research-type micropipettes (single-channel or multi-channel).
  • Practical wall mounting frees up space on the lab work bench
  • Compatible with adapters for Multipette® plus and Varipette® micropipettes