Tlačiť
Safety spectacles, uvex astrospec
Dodávateľ: Uvex
Astrospec
111-1575EA 28.84 EUR
111-1575 111-8032 111-8003 111-8004 111-8002
Safety spectacles, uvex astrospec
Brýle
These modern safety glasses are available in a range of colours and styles. They all feature a high impact-resistant polycarbonate lens with integrated side protectors.
  • Duo-flex side arms with 4-stage side arm length adjustment and lens inclination allows wearers to achieve individual pressure free fit
  • Temple pads for comfort and better fit
  • Special coating allows easy cleaning
  • Simple lens replacement
Weight: 18 g
Certifikácie: EN 166, EN 170