Tlačiť
Safety spectacles, B-Line
Dodávateľ: Bollé Safety
111-0978EA 6.44 EUR
BOLEBL10CI
Safety spectacles, B-Line
Single lens safety glasses offering a panoramic field of vision. The frame is manufactured from nylon.
  • Integrated side shields
  • With headband
Weight: 30 g
Certifikácie: EN 166, EN 170