Tlačiť
Stoppers, conical, Iso-Versinic®
Dodávateľ: Saint-Gobain
Iso-Versinic®
217-0885EA 217.43 EUR
217-0885
Stoppers, conical, Iso-Versinic®
Zátky Zátky kaučukové (gumové)
Made of Fluoroelastomer Viton®. Excellent chemical resistance, even at high temperatures.
  • Resistant to many acids and aggressive chemicals even in high concentrations and at high temperatures, particularly hydrocarbons, oils and strong acids, aromatic and chlorinated solvents
  • Convenient pack sizes
  • Temperature resistant from −20 to +200 °C (up to +300 °C for short periods)