Chcete sa opäť prihlásiť?
Laboratory trays
Dodávateľ: Kartell
plastilab®
PVC, white.
  • Thick walled and robust
  • Heat resistant up to +70 °C
  • Smooth surfaces allow easy cleaning
With ribs on base. Highly versatile trays, ideal for a wide range of applications. The ribs on the base make these trays ideal for use as developing trays.
Log in to see your contract pricing and availability.
  Zapamätať si ma na tomto zariadení
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Š × H × V Pk Číslo v katalógu VWR Množstvo
200×150×45 1 216-2010
250×200×60 1 216-2011
320×260×70 1 216-2012
350×300×80 1 216-2013
430×330×90 1 216-2014
520×420×90 1 216-2020
540×420×180 1 216-2030
675×530×100 1 216-2025
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo