Tlačiť
Dollys, Nalgene®
Dodávateľ: Thermo Fisher Scientific
129-0178EA 1562.58 EUR
129-0178
Dollys, Nalgene®
Vozíky
For transporting round tanks, carboys and balloon flasks as well as other round containers with up to 521 mm Ø.
  • Resistant to corrosion and to acids and alkalis
  • Strong, maximum load capacity 227,3 kg
  • Casters designed to minimise stress on flooring
For transporting round tanks, carboys and balloon flasks as well as other round containers with up to 521 mm Ø.
  • Resistant to corrosion and to acids and alkalis
  • Strong, maximum load capacity 227,3 kg
  • Casters designed to minimise stress on flooring
Stainless steel, rustproof.