Tlačiť
Microcentrifuge tubes, Safe-Lock PCR clean, 0,5/1,5/2,0 ml, Eppendorf®
Dodávateľ: Eppendorf
Safe-Lock PCR clean
211-2165EA 68.02 EUR
211-2165 211-2164 211-2163
Microcentrifuge tubes, Safe-Lock PCR clean, 0,5/1,5/2,0 ml, Eppendorf®
Zkumavky Mikrozkumavky centrifugační
PP, graduated, with attached cap, autoclavable when open.
  • Can withstand centrifugation up to 30000×g for 0,5 and 1,5 ml and up to 25000×g for 2,0 ml
  • The hook on the patented hinged lid clips around the rim of the test tube, preventing the tube from opening accidentally
  • Free of human DNA, DNase, RNase and PCR inhibitors (PCR clean)