Tlačiť
Microcentrifuge tubes, Microcentrifuge Tubes, 0,5/1,5/2,0 ml, Safe-Lock, Eppendorf®
Dodávateľ: Eppendorf
Safe-Lock
211-2145EA 65.2 EUR
211-2145 211-2123 211-2134 211-2144 211-2133 211-2122 211-2132 211-2121 211-2143 211-2142 211-2131 211-2120 211-2149 211-2125 211-2124 211-2135 211-2141 211-2130 211-2140 211-2129 211-2139
Microcentrifuge tubes, Microcentrifuge Tubes, 0,5/1,5/2,0 ml, Safe-Lock, Eppendorf®
Zkumavky Mikrozkumavky centrifugační
PP, graduated, with attached cap, autoclavable when open.
  • Can withstand centrifugation up to 30000×g for 0,5 and 1,5 ml and up to 25000×g for 2,0 ml
  • Frosted writing surface, easily opened using one hand
  • The hook on the patented hinged lid clips around the rim of the test tube, preventing the tube from opening accidentally
Micropestle for microcentrifuge tubes
White plastic. For resuspension of pellets in 1,5 and 2,0 ml microtubes
Simple and easy handling
Good chemical resistance
High thermal resistance, autoclavable