Tlačiť
Pressure tubing, Tygon® S3™ B-44-4X I.B.
Dodávateľ: Saint-Gobain
228-1019EA 0 EUR
228-1019
Pressure tubing, Tygon® S3™ B-44-4X I.B.
Hadičky Přenosová hadička
Tygon® tubing, transparent, with fabric lining.
  • Clear wall for visualisation of flow
  • Pressure resistance is four times higher than that of non reinforced tubing
  • Non toxic, taste-free and odourless
This pressure tubing ensures rapid and simple start-up due to its unique flexibility. This means shorter tube lengths, fewer connections and less effort than with rigid systems. Includes a fabric lining made of strong polyester braid for high pressure resistance. The smooth inner walls of the tubing do not trap substances containing bacteria so this tubing is ideally suited to applications in the food and beverage industry.
Offers excellent resistance to the alkaline degreasing agents.
Temperature resistance: −44 to +74 °C
Hardness (Shore durometer A): 65
Density: 1,21
Certifikácie: Complies with the requirements of the US Food and Drugs Agency (FDA).