Tlačiť
SP Bel-Art weight strips
Dodávateľ: Bel-Art
214-1311EA 85.76 EUR
BELAF183210212
SP Bel-Art weight strips
Soft lead sheet coated with flexible Vikem® vinyl has many uses for stabilizing laboratory equipment in water baths or any application where extra weight is needed.
  • Sheets may be bent into cylinders, 'S' curves, 'U' curves or twists to hold down various container types
  • Vikem® vinyl coating prevents the lead from leaving marks on equipment
  • Coated lead sheets are 2,2 mm thick