Tlačiť
Dual-gel electrophoresis system, Owl™ P9DS
Dodávateľ: Thermo Fisher Scientific
700-2350EA 2748.15 EUR
700-2350 730-0318 700-2351
Dual-gel electrophoresis system, Owl™ P9DS
Systémy elektroforetické Systémy pro vertikální gelovou elektroforézu
The Owl™ P9DS dual gel system is designed for quick and easy casting and running of one or two gels on the same device. When used with the available combs, spacers, alumina plates and glass, this system is an ideal tool for SDS electrophoresis and 2-D electrophoresis.
  • Simple clamping mechanism makes loading glass cassettes, and most precast gels, trouble free
Gel W×H: 160×140 to 160 mm
Footprint W×H×D: 250×195×130 mm
Running buffer volume: 550 ml (upper chamber 200 ml, lower chamber 350 ml)
Informace o dodávce: Complete system includes upper buffer chamber with internal cooling core, lower buffer chamber, SuperSafe lid with attached power leads, 4 blank glass plates, 4 notched glass plates, 4 combs 1,5 mm thick (2×10 well, 2×15 well), spacer set (1,5 mm thick), spacer placer, and blocking plate for single gel operation.