Tlačiť
Mini-vertical blotting system (4-place), EBX-700
Dodávateľ: CBS Scientific
CBSCEBX-700
Mini-vertical blotting system (4-place), EBX-700
This mini-vertical blotting system can accommodate gels of dimensions 100×100, 100×90, or 100×80 mm and is capable of transferring 4 gels simultaneously under identical temperature controlled buffer conditions. The system is designed for Western blotting.
  • Freezer blocks fit into compartments in the lower reservoir for cooling, eliminating the need for an external cooling apparatus
  • Colour coded anode and cathode for additional safety
  • Seamless moulded tank design with space for a magnetic stirring bar
  • Uses less than 1 litre of buffer (cathode reservoir: 190 ml; anode reservoir: 750 ml)
Informace o dodávce: Each unit includes 4× blotting cassettes with sponge pads, 4× freezer blocks, lower reservoir with 4× gel capacity, and safety cover with attached leads.