Tlačiť
Disinfection system, Minncare® Mini Dry Fog™
Dodávateľ: Minntech
Mini Dry Fog®
141-0045
Disinfection system, Minncare® Mini Dry Fog™
Systémy dezinfekční
Mini Dry Fog® is a simple, economical system for disinfecting small volumes (1 to 20 m³) with airborne agents. It replaces procedures that use formol or formaldehyde and can be used to disinfect gowning or decontamination areas, confined spaces and enclosed areas with laminar airflow (biohazard). All that is needed for 90 to 120 minutes (max.) disinfection is a compressed air connection (5 bar). Mini Dry Fog® generates a dry fog that does not wet surfaces and a vapour phase with proven sporicidal effectiveness (see Minncare Dry Fog™ system).
  • Effective, with proven sporicidal efficacy greater than 6 log
  • Monitoring and traceability: supplied together with software for optimising procedures and validating disinfections
  • fog evaporates and leaves no residue on surfaces
  • Documented in accordance with pharmaceutical standards
Certifikácie: AFNOR NF-T72-381
Conforms to recommended procedures: Satisfies the latest FDA recommendations for use of sporicides in aseptic areas.