Tlačiť
Post-column derivatisation system, Pinnacle
Dodávateľ: Pinnacle
903-0080 903-0081
Post-column derivatisation system, Pinnacle
Systémy chromatografické Systémy derivatizační
Optimised for analysing amino acids, carbamates, mycotoxins, antibiotics, and many other applications. Post-column derivatisation technology improves the sensitivity and/or selectivity of the HPLC chromatography technique as applied in a wide range of procedures.
  • Programming the column oven temperature gradient improves separation and the cycle duration
  • Electronic syringe pump for continuous, regular flow
  • One-piece, ceramic pump
  • Electronic valves
  • Reaction cartridge are easily replaceable
  • Column oven uses circulating air for more efficient heating and cooling
PCX control software enables the introduction of reagent to be controlled precisely.