Tlačiť
Modular heating mantles with stirrer, Electromantle® OMCA series
Dodávateľ: Electrothermal
Electromantle®
453-0165EA 4574.92 EUR
453-0165 453-0091 453-0090
Modular heating mantles with stirrer, Electromantle® OMCA series
Ohřívače Hnízda topná
Heater element temperature max. 220 °C; magnetic stirrer speed: 100 to 800 min⁻¹. Base unit accommodates choice of three interchangeable cartridges: 250, 500 or 1000 ml round bottom flask sizes.
  • Constructed of ABS flame-retardant thermoplastic
  • Heater cartridge case is spun aluminium with polyester paint coating
Ceramic fibre thermal insulation permits faster heat up while reducing energy consumption.
Three digit display toggles between set temperature and actual temperature ±1 °C.