Tlačiť
Stereomikroskopy, K series
Dodávateľ: Motic
MOTI1100200900211
Stereomikroskopy, K series
Mikroskopy
The K series provides distortion-free 3-D images for biological and industrial inspection.
  • Fatigue-free viewing
  • Various contrast and documentation options
  • Common Main Objective (CMO) optical system
Informace o dodávce: Supplied with frosted glass (except 630-0139), black and white stages and dust cover.