Tlačiť
Multi-frequency ultrasonic baths, Transsonic TI-H series
Dodávateľ: Elma
142-0155EA 3163.59 EUR
ELMA2390400010
Multi-frequency ultrasonic baths, Transsonic TI-H series
The Transsonic TI-H series comes in different sizes with 25 to 45 kHz. The ergonomic design and superior material compatibility are intended for industrial use in production, repair shops and service centres. These ultrasonic baths are equipped with timer, variable from 0 to 15 minutes.
  • Two switchable frequencies for intensive and gentle cleaning
  • Vibration tanks made of special stainless steel for heavy-duty use and long service life
  • Degas: For fast, effective degassing of the cleaning fluid
  • Sweep: For optimum sound field distribution
  • Thermostatically adjustable heating (30 to 80 °C)
With two switchable frequencies.
Objednávacie informácie: Other models are available, please contact us for further information.