Tlačiť
Poly-D-lysine/laminin culture dishes, Corning® BioCoat™
Dodávateľ: Corning
Corning® BioCoat®
734-0140EA 313.88 EUR
734-0140
Poly-D-lysine/laminin culture dishes, Corning® BioCoat™
Misky kultivační Misky pro tkáňové kultury
PS coated with a uniform application of poly-D-lysine and laminin. Promotes neuronal cell attachment and differentiation for a variety of applications.