Tlačiť
Nálevky
Dodávateľ: Pall Laboratory
PALL4240
Nálevky
Stainless steel construction offers excellent chemical resistance.
  • Threaded collar seals to base
  • Autoclavable
  • Connection: Stem inserts into standard size No. 2 rubber stopper
Vacuum filtration of small volumes of solutions for particulate contamination analysis.