Tlačiť
Temperature loggers, Kit T 350/220
Dodávateľ: Binder
BIND8012-0714EA 1324.55 EUR
BIND8012-0714 BIND8012-0715
Temperature loggers, Kit T 350/220
This data logger kit can record temperature.
  • Power supply: 3,6 volt lithium replaceable battery with status display
  • Calibration certificate included
Contains useful accessories for mounting the logger on the unit, including cable bushings and a sensor mounting bracket.
Ambient conditions: −35 to +55 °C.