Tlačiť
Orbital shakers, Corning® LSE™
Dodávateľ: Corning
Corning® LSE™
CORN6781-NPEA 2082.41 EUR
CORN6781-NP CORN6782-NP
Orbital shakers, Corning® LSE™
Třepačky Třepačky orbitální
The Corning LSE™ orbital shaker is suitable for a variety of shaking and mixing applications. A powerful brushless motor and counter-balanced shaking mechanism provide smooth operation for years of trouble-free use. Different platforms (order separately) are available and are easily interchangeable.
  • Two stainless steel platform options
  • 19 mm orbit for aeration of cultures and staining gels
  • Large digital display with single-knob control
  • Digital control of shaking speed and time
  • Safe for cold room or incubator use
Objednávacie informácie: Supplied without platform, different platforms are available separately as optional accessories.