Tlačiť
Goggles, Maxim™ Hybrid
Dodávateľ: 3M
MMMAMAXHYBEA 0 EUR
MMMAMAXHYB
Goggles, Maxim™ Hybrid
The Maxim™ Hybrid goggle is a very lightweight, low-profile form of eyewear protection that provides high comfort and unsurpassed protection against liquid droplets and mechanical hazards. The slim design offers improved comfort and wearer acceptance. The comfortable nylon head strap is fully adjustable. The DX-coating of the lens resists fogging, scratching and chemicals.
  • High performance ventilation system overcomes fogging problems
  • Patented aspherical lens for 180° field of view and superior eye coverage
  • Special design for improved compatibility with other PPE
Weight: 28 g
Certifikácie: EN 166