Tlačiť
Evaporating dishes, PFA
Dodávateľ: Saint-Gobain
VERND1069543EA 165.99 EUR
VERND1069543
Evaporating dishes, PFA
Made of PFA, translucent.
  • Chemically inert
  • Perfect for microwave use
  • Smooth, non-stick surface
Ideal for trace metal analysis.