Tlačiť
Bottles, narrow neck, round, PFA, with screw caps, Chemware®
Dodávateľ: Saint-Gobain
VERND1069068EA 272.82 EUR
VERND1069068
Bottles, narrow neck, round, PFA, with screw caps, Chemware®
PFA, translucent.
  • Maximum chemical resistance
  • Heat resistant from -200 to +260 °C, autoclavable
  • Easy to clean