Tlačiť
Torque wrenches, Nalgene®
Dodávateľ: Thermo Fisher Scientific
Nalgene®
NALG2195-6177EA 874.72 EUR
NALG2195-6177
Torque wrenches, Nalgene®
Klíče
These wrenches can be used to confirm capping machine settings, on-line inspection and to check torques.
  • Ensure leakproof sealing of closures
Objednávacie informácie: Order fittings that match closure resin.