Tlačiť
Accessories for laboratory atting Chemware®
Dodávateľ: Saint-Gobain
VERND1069138EA 284.32 EUR
VERND1069138 VERND1069140
Accessories for laboratory atting Chemware®
The chemically inert PTFE cord can be used to create a dipping basket for corrosive baths. Therefore, the edges of the Chemware® PTFE laboratory matting need to be secured. The cord then acts as a 'thread'.
  • Corrosion resistant
  • Chemically inert
  • Easy to tie