Tlačiť
Forceps, Chemware®
Dodávateľ: Saint-Gobain
Chemware®
VERND1069095EA 1135.95 EUR
VERND1069095
Forceps, Chemware®
Pinzety
These forceps are made from PTFE resin with an encapsulated metal insert for strength. Ideal for laboratory work involving hot, cold, delicate or highly reactive materials.
  • Chemically inert
  • Non contaminating
  • Will not scratch delicate surfaces