Tlačiť
Funnels, Chemware®
Dodávateľ: Saint-Gobain
VERND1069642EA 427.75 EUR
VERND1069642
Funnels, Chemware®
These PFA Chemware® funnels are non-contaminating, inert with low surface energy facilitates quantitative transfer.
  • Very good chemical resistance
  • Very good temperature resistance
  • Easy to clean