Tlačiť
Acid transfer tubing, UV resistant, Versilon™ R-3400
Dodávateľ: Saint Gobain Performance Plastics
VERNAAE00005EA 0 EUR
VERNAAE00005
Acid transfer tubing, UV resistant, Versilon™ R-3400
Hadičky Přenosová hadička
Black.
  • Resistant to a broad range of chemicals including strong acids, bleach, and glycols
  • Black tubing is also resistant to ultraviolet light, ozone and weathering
  • Abrasion-resistant
Flexible tubing is ideal for permanent or temporary chemical installations. Can be used in the transport of semi-paste adhesives, liquid fertilizers and pesticides, as well as general production and maintenance.
Hardness (Shore durometer A): 64
Temperature resistance: +74 °C
Density: 1,31