Tlačiť
Water baths, SWB series
Dodávateľ: Cole Parmer
stuart®
STUASWB15DEA 1238.94 EUR
STUASWB15D STUASWB6D STUASWB24D
Water baths, SWB series
Lázně Lázně vodní, nemíchané
Digital water bath with stainless steel tank and sturdy metal outer casing, heating element which facilitates rapid warm-up, electronic control ensures temperature stability, and the selection of temperature is easy and accurate with LCD display.
  • Digital set and display of bath temperature
  • Adjustable safety thermostat
  • Corrosion resistant tank
  • Choice of three sizes: 6, 15 and 24 L
Objednávacie informácie: Bath cover and lid are also available and can be ordered separately.