Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

1-Methylcycloheptanol ≥98%

Dodávateľ: Alfa Aesar
Pozor

Purity: 98%
CAS No.: 3761-94-2
Molecular Formula: C8H16O
Formula Weight: 128.22
Refractive Index: 1.4720

Vzorec: C₈H₁₆O
MW: 128,21 g/mol
Teplota varu: 183,5 °C (1013 hPa)
Teplota tavenia: -4 °C
Hustota: 0,918 g/cm³ (20 °C)
Číslo CAS: 3761-94-2
EINECS: 223-179-9

Log in to see your contract pricing and availability.
  Zapamätať si ma na tomto zariadení
 

OBJEDNÁVKA

Rozšíriť všetko / Schovať všetko
Pk Číslo v katalógu VWR Množstvo
25 g B20393.14
5 g B20393.06
Rozšíriť všetko / Schovať všetko
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ŠPECIFIKÁCIE

Formula weight 128.22
CAS No. 3761-94-2
Purity 98%
Refractive Index 1.4720
Molecular Formula C8H16O
MDL No. MFCD00021672
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo