Tlačiť

ALFAB20792

ALFAB20792.06
ALFAB20792
Purity: 98%
CAS No.: 7731-28-4
Molecular Formula: C7H14O
Formula Weight: 114.19
Flash Point: 63°(145°F)
Refractive Index: 1.4610
Melting Point: -10°
Boiling Point: 172-173°
MDL No.: MFCD00064170
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR