Tlačiť
UV/Vis spectrophotometers, Ultrospec™ 8000
Dodávateľ: Biochrom
Ultrospec™
634-0888EA 0 EUR
634-0888 634-0886 634-0887
UV/Vis spectrophotometers, Ultrospec™ 8000
Spektrofotometry a fotometry Spektrofotometry UV/Vis
UV/Vis spectrophotometers designed for analyses in controlled or regulated laboratories. Stand-alone or PC controlled versions are available for both series.
  • Full European Pharmacopoeia compatibility
  • Colour touch screen for stand-alone models only
  • Equation editor software for custom calculations to be applied to measured data
  • Multi-user analytical instrument with secure login to protect methods
  • Software for wavelength scanning, fixed wavelength, standard curves, kinetics and prestored life science methods
Objednávacie informácie: For 21 CFR part 11 compatibility, optional Datrys™ CFR software and IQ/OQ system qualification can also be made available. For details of the full range of accessories, please contact your local Avantor sales office.