Chcete sa opäť prihlásiť?
Accessories for fraction collector, SuperFrac™
Dodávateľ: GE Healthcare
SuperFrac™
Log in to see your contract pricing and availability.
  Zapamätať si ma na tomto zariadení
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Opis Pk Číslo v katalógu VWR Množstvo
Tube racks, type A each for 26 tubes 12 - 13 mm 12 18-1010-20
Tube racks, type B each for 20 tubes 16 - 18 mm 9 18-1010-21
Tube racks, type C each for 14 scintillation vials 28 mm 6 18-1010-22
Rack tray 1 80-1071-43
HiFlow rack kit including 4 HiFlow rack funnels, HiFlow nozzle, drop nozzle 2,7, tube holder, 2 stoppers, 20 m silicone tubing 8 mm i.d. 12 mm o.d., 2 m silicone tubing 4 mm i.d. 6 mm o.d., 2 m S-flanged tubing, Allen key, 2 side plates and 2 shafts 1 KIT 18-5000-01
Cable, Fisher 1 80-1266-31 Blocked
Blocked Vaša organizácia zablokovala tento produkt. Prosím kontaktujte oddelenie nákupu pre viac informácii.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo