Chcete sa opäť prihlásiť?
Jars, with ground joint and screw cap
Dodávateľ: BOLA
BOLA
Jars, made of PFA, transparent with screw-cap and integrated ground joints made of PTFE.
  • Good chemical resistance
  • Temperature resistance from −200 to +250 °C
Suitable for used as an evaporation vessel or reaction vessel for rotary evaporators.
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Kapacita Výška Vonkajší Ø. Pk Číslo v katalógu VWR Množstvo
1000 ml 193 mm 107 mm 1 BOHLA159-24
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo