Chcete sa opäť prihlásiť?
Ergonomic mats, basic bubble and basic smooth
Dodávateľ: VWR Collection
VWR®
These mats are made of low density polyurethane providing an ergonomic, anti-fatigue profile. Bubble mats are designed for dry, non critical environments, reducing trip hazards, body aches, quality issues and sick leave. The smooth, flat mats are ideal for dry, non critical environments where sitting and standing work areas are combined, or where carts or trolleys must be used.
  • Moulded bevelled edges will not curl up, thus eliminating trip hazards
  • Any size or any shape - will always be one piece: No overlapping or worn out connecting mats
  • Ergonomic design stimulates the body to increase blood circulation and alertness
  • Silicone- and latex-free
  • Warranty up to 3 years
Certifikace: DIN 53505, EN ISO 527-1, ASTM D 624
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Basic Bubble
D × Š Hrúbka Farba Pk Číslo v katalógu VWR Množstvo
1200×1200 mm 15 mm Grey 1 115-1185
1200×1500 mm 15 mm Grey 1 115-1186
1200×1800 mm 15 mm Grey 1 115-1187
1200×2100 mm 15 mm Grey 1 115-1188
1200×2400 mm 15 mm Grey 1 115-1189
1200×2700 mm 15 mm Grey 1 115-1190
1200×3000 mm 15 mm Grey 1 115-1191
1200×3300 mm 15 mm Grey 1 115-1192
1200×3600 mm 15 mm Grey 1 115-1193
1200×4200 mm 15 mm Grey 1 115-1195
1200×4500 mm 15 mm Grey 1 115-1196
1200×4800 mm 15 mm Grey 1 115-1197
1200×5100 mm 15 mm Grey 1 115-1198
1200×5400 mm 15 mm Grey 1 115-1199
1200×5700 mm 15 mm Grey 1 115-1200
1200×6000 mm 15 mm Grey 1 115-1201
600×1200 mm 15 mm Grey 1 115-1150
600×1500 mm 15 mm Grey 1 115-1151
600×1800 mm 15 mm Grey 1 115-1152
600×2100 mm 15 mm Grey 1 115-1153
600×2400 mm 15 mm Grey 1 115-1154
600×2700 mm 15 mm Grey 1 115-1155
600×3000 mm 15 mm Grey 1 115-1156
600×3300 mm 15 mm Grey 1 115-1157
600×3600 mm 15 mm Grey 1 115-1158
600×3900 mm 15 mm Grey 1 115-1159
600×4200 mm 15 mm Grey 1 115-1160
600×4500 mm 15 mm Grey 1 115-1161
600×5100 mm 15 mm Grey 1 115-1163
600×5400 mm 15 mm Grey 1 115-1164
600×5700 mm 15 mm Grey 1 115-1165
600×6000 mm 15 mm Grey 1 115-1166
600×900 mm 15 mm Grey 1 115-1149
900×1200 mm 15 mm Grey 1 115-1168
900×1500 mm 15 mm Grey 1 115-1169
900×1800 mm 15 mm Grey 1 115-1170
900×2100 mm 15 mm Grey 1 115-1171
900×2400 mm 15 mm Grey 1 115-1172
900×2700 mm 15 mm Grey 1 115-1173
900×3000 mm 15 mm Grey 1 115-1174
900×3300 mm 15 mm Grey 1 115-1175
900×3600 mm 15 mm Grey 1 115-1176
900×3900 mm 15 mm Grey 1 115-1177
900×4200 mm 15 mm Grey 1 115-1178
900×4500 mm 15 mm Grey 1 115-1179
900×4800 mm 15 mm Grey 1 115-1180
900×5100 mm 15 mm Grey 1 115-1181
900×5400 mm 15 mm Grey 1 115-1182
900×5700 mm 15 mm Grey 1 115-1183
900×6000 mm 15 mm Grey 1 115-1184
900×900 mm 15 mm Grey 1 115-1167
Basic Smooth
D × Š Hrúbka Farba Pk Číslo v katalógu VWR Množstvo
1200×1200 mm 11 mm Grey 1 115-1238
1200×1500 mm 11 mm Grey 1 115-1239
1200×1800 mm 11 mm Grey 1 115-1240
1200×1800 mm 11 mm Grey 1 115-1249
1200×1800 mm 11 mm Grey 1 115-1255
1200×2100 mm 11 mm Grey 1 115-1241
1200×2400 mm 11 mm Grey 1 115-1242
1200×2700 mm 11 mm Grey 1 115-1243
1200×3000 mm 11 mm Grey 1 115-1244
1200×3300 mm 11 mm Grey 1 115-1245
1200×3600 mm 11 mm Grey 1 115-1246
1200×3900 mm 11 mm Grey 1 115-1247
1200×4200 mm 11 mm Grey 1 115-1248
1200×4800 mm 11 mm Grey 1 115-1254
1200×5400 mm 11 mm Grey 1 115-1256
1200×5700 mm 11 mm Grey 1 115-1257
1200×6000 mm 11 mm Grey 1 115-1258
600×1200 mm 11 mm Grey 1 115-1203
600×1500 mm 11 mm Grey 1 115-1204
600×1800 mm 11 mm Grey 1 115-1205
600×2100 mm 11 mm Grey 1 115-1206
600×2400 mm 11 mm Grey 1 115-1207
600×2700 mm 11 mm Grey 1 115-1208
600×3000 mm 11 mm Grey 1 115-1209
600×3300 mm 11 mm Grey 1 115-1210
600×3600 mm 11 mm Grey 1 115-1211
600×3900 mm 11 mm Grey 1 115-1212
600×4500 mm 11 mm Grey 1 115-1214
600×4800 mm 11 mm Grey 1 115-1215
600×5100 mm 11 mm Grey 1 115-1216
600×5400 mm 11 mm Grey 1 115-1217
600×5700 mm 11 mm Grey 1 115-1218
600×6000 mm 11 mm Grey 1 115-1219
600×900 mm 11 mm Grey 1 115-1202
900×1200 mm 11 mm Grey 1 115-1221
900×1500 mm 11 mm Grey 1 115-1222
900×1800 mm 11 mm Grey 1 115-1223
900×2100 mm 11 mm Grey 1 115-1224
900×2400 mm 11 mm Grey 1 115-1225
900×2700 mm 11 mm Grey 1 115-1226
900×3000 mm 11 mm Grey 1 115-1227
900×3300 mm 11 mm Grey 1 115-1228
900×3600 mm 11 mm Grey 1 115-1229
900×3900 mm 11 mm Grey 1 115-1230
900×4200 mm 11 mm Grey 1 115-1231
900×4500 mm 11 mm Grey 1 115-1232
900×4800 mm 11 mm Grey 1 115-1233
900×5100 mm 11 mm Grey 1 115-1234
900×5400 mm 11 mm Grey 1 115-1235
900×5700 mm 11 mm Grey 1 115-1236
900×6000 mm 11 mm Grey 1 115-1237
900×900 mm 11 mm Grey 1 115-1220
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo