Chcete sa opäť prihlásiť?
Accessories
Dodávateľ: Syngene (a division of the Synoptics group)
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Opis Pk Číslo v katalógu VWR Množstvo
UV to white light converter screen, 305×330 mm 1 730-1396
UV to white light converter screen, 200×200 mm 1 730-1395
UV to blue light conversion screen 210×260 mm 1 730-1493
UV to blue light conversion screen 250×300 mm 1 730-1494
White light pad, fold down, with brackets, 200×140 mm 1 730-1391
Epi UV module, with 2×365 nm tubes 1 730-1392
Epi UV module, with 2×254 nm tubes 1 730-1394
Epi UV module, with 1×302 nm and 1×365 nm tubes 1 733-2304
Epi UV module, with 1×365 nm and 1×254 nm tubes 1 SYNOEPI-MOD-SW-LW
Epi UV module, with 1×302 nm and 1×254 nm tubes 1 SYNOEPI-MOD-MW-SW
Epi UV module, with 2×302 nm tubes 2 733-2615
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo