Tlačiť
Stoppers for flasks with ground joints, SP Bel-Art Safe-lab™
Dodávateľ: Bel-Art
BELAF201961502EA 228.31 EUR
BELAF201961502
Stoppers for flasks with ground joints, SP Bel-Art Safe-lab™
Zátky Zátky PE/LDPE/HDPE
Use with boiling and distilling flaks. Safe-Lab™ stoppers are easily removed from standard tapered glassware by turning the red extraction nut to apply a safe, gentle and even force against the mouth of the vessel, helping to eleminate accidents and breakage.
  • PTFE, Solid Teflon®
  • Avoid risking injury with dangerous removal techniques
  • Avoid tapping glassware against a bench top
  • Autoclavable
Use with boiling and distilling flaks. Safe-lab™ stoppers are easily removed from standard tapered glassware by turning the red extraction nut to apply a safe, gentle and even force against the mouth of the vessel, helping to eleminate accidents and breakage.
  • Avoid risking injury with dangerous removal techniques
  • Avoid tapping glassware against a bench top
  • Autoclavable