Tlačiť
Cooling plate, Iceberg™
Dodávateľ: CellPath
Iceberg™
720-0016 720-2349
Cooling plate, Iceberg™
Chladiče Chladiče stolní
These cooling plate are kept in a freezer or cryostat and placed in the tray to cool tissue blocks. The plate can be cooled to –20 °C and keeps tissue cool for up to four hours.
  • Can cool up to 28 tissue blocks simultaneously
  • The tray collects condensation
  • From black aluminium, tray from moulded plastic
A cooling plate is used to keep the tissue in good condition when it is processed into coupes.